Skip to Content
Merck
All Photos(2)

Key Documents

23-034-M

Sigma-Aldrich

p53 Protein, 10 µg

Full length, recombinant human p53. N-terminal c-Myc, 6His-tagged. For use in Enzyme Assays. Functions as a substrate in ubiquitination assays.

Synonym(s):

Cellular tumour antigen p53, Antigen NY-CO-13, Phosphoprotein p53, Tumour suppressor p53, TP53

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€682.00
250 μG
€3,950.00

€682.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€682.00
250 μG
€3,950.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€682.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

solubility

water: soluble

GenBank accession no.

storage temp.

−70°C

Gene Information

General description

Research area: Apoptosis

N-terminal GST-tagged, recombinant human TRAF6 full length, expressed in E.coli. The p53 protein is a tumor suppressor encoded by TP53 gene [1] located on chromosome 17.[2]

Application

Ubiquitination Cascade Comoponent: Substrate

Biochem/physiol Actions

p53 is expressed in a wide variety of tissues and plays a major role in preventing tumour development. It is involved in cell cycle regulation, functioning as a trans-activator to negatively regulate cell division by controlling a set of genes required for this process. It acts as a tumour suppressor in many tumour types by stimulating either apoptosis or growth arrest, depending on the physiological circumstances and the cell type. Defects in p53 expression have been implicated in several diseases, including choroid plexus papilloma, lung cancer, head/neck squamous cell carcinomas, oesophageal squamous cell carcinoma, Li-Fraumeni syndrome, and hereditary adrenocortical carcinoma.

Physical form

Purified using immobilized metal affinity chromatography.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hany E Marei et al.
Cancer cell international, 21(1), 703-703 (2021-12-26)
The p53 protein is a transcription factor known as the "guardian of the genome" because of its critical function in preserving genomic integrity. The TP53 gene is mutated in approximately half of all human malignancies, including those of the breast
The p53 tumor suppressor protein
Genes & diseases (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service