Skip to Content
Merck
All Photos(1)

Documents

219674

Sigma-Aldrich

CFTR Inhibitor-172

InSolution, ≥95%

Synonym(s):

InSolution CFTR Inhibitor-172, (Z)-4-((4-Oxo-2-thioxo-3-(3-(trifluoromethyl)phenyl)thiazolidin-5-ylidene)methyl)benzoic acid, CFTRinh-172 in DMSO, 3-((3-Trifluoromethyl)phenyl)-5-((4-carboxyphenyl)methylene)-2-thioxo-4-thiazolidinone, 3-((3-Trifluoromethyl)phenyl)-5-((4-carboxyphenyl)methylene)-2-thioxo-4-thiazolidinone, (Z)-4-((4-Oxo-2-thioxo-3-(3-(trifluoromethyl)phenyl)thiazolidin-5-ylidene)methyl)benzoic acid, CFTRinh-172 in DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10F3NO3S2
CAS Number:
Molecular Weight:
409.40
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

shipped in

dry ice

storage temp.

−70°C

InChI

1S/C18H10F3NO3S2/c19-18(20,21)12-2-1-3-13(9-12)22-15(23)14(27-17(22)26)8-10-4-6-11(7-5-10)16(24)25/h1-9H,(H,24,25)/b14-8-

InChI key

JIMHYXZZCWVCMI-ZSOIEALJSA-N

General description

A cell-permeable 2-thio-4-thiazolidinone compound that acts as a potent, reversible, rapid, and voltage-independent inhibitor of CFTR (cystic fibrosis transmembrane conductance regulator)-mediated Cl- transport in human airway cells (Ki ~ 300 nM). It does not prevent elevation of cellular cAMP, nor does it inhibit non-CFTR Cl- channels, MDR-1, ATP-sensitive K+ channels, or a series of other transporters even at concentrations as high as 5 µM. Shown to block cholera toxin-induced intestinal fluid secretion in mice. May be also useful for reducing intestinal fluid loss in other secretory diarrheas. The solid form of this compound (Cat. No. 219670) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CFTR ion-channel
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/244 µL) solution of CFTR Inhibitor-172 (Cat. No. 219670) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (70°C). Aliquots are stable for up to 3 months at -70°C.

Other Notes

Yang, H., et al. 2003. J. Biol. Chem.278, 35079.
Al-Awqati, Q., et al. 2002. J. Clin. Invest.110, 1599.
Ma, T., et al. 2002. J. Clin. Invest.110, 1651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service