Skip to Content
Merck
All Photos(1)

Key Documents

219361

Sigma-Aldrich

Anti-Cathepsin D Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€553.00

€553.00


Estimated to ship on28 April 2025Details



Select a Size

Change View
1 ML
€553.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€553.00


Estimated to ship on28 April 2025Details


biological source

rabbit

Quality Level

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-Cathepsin D, rabbit polyclonal, recognizes the ~50 kDa latent and the ~30 kDa active forms in breast carcinoma cell line. It is validated for ELISA, Western blotting, and immunoelectrophoresis.
Rabbit polyclonal antibody purified by ammonium sulfate precipitation. Recognizes the ~50 kDa latent and the ~30 kDa active forms of cathepsin D.
Recognizes the ~50 kDa latent and the ~30 kDa active forms of cathepsin D in breast carcinoma cell line.

Immunogen

Human
purified human liver cathepsin D

Application

ELISA (≥1:1000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles.

Analysis Note

Positive Control
Human liver tissue

Other Notes

Monospecific for cathepsin D as determined by a single arc by immunoelectrophoresis against crude live extract and purified cathepsin D. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Heinrich et al.
Cell death and differentiation, 11(5), 550-563 (2004-01-24)
Acidic noncaspase proteases-like cathepsins have been introduced as novel mediators of apoptosis. A clear role for these proteases and the acidic endolysosomal compartment in apoptotic signalling is not yet defined. To understand the role and significance of noncaspases in promoting
Rachel Allison et al.
The Journal of cell biology, 216(5), 1337-1355 (2017-04-09)
Contacts between endosomes and the endoplasmic reticulum (ER) promote endosomal tubule fission, but the mechanisms involved and consequences of tubule fission failure are incompletely understood. We found that interaction between the microtubule-severing enzyme spastin and the ESCRT protein IST1 at
Maya M Polovitskaya et al.
American journal of human genetics, 107(6), 1062-1077 (2020-11-21)
Dysfunction of the endolysosomal system is often associated with neurodegenerative disease because postmitotic neurons are particularly reliant on the elimination of intracellular aggregates. Adequate function of endosomes and lysosomes requires finely tuned luminal ion homeostasis and transmembrane ion fluxes. Endolysosomal
Gonzalo A Mardones et al.
Molecular biology of the cell, 18(9), 3486-3501 (2007-06-29)
The sorting of acid hydrolase precursors at the trans-Golgi network (TGN) is mediated by binding to mannose 6-phosphate receptors (MPRs) and subsequent capture of the hydrolase-MPR complexes into clathrin-coated vesicles or transport carriers (TCs) destined for delivery to endosomes. This
Daniel J Metcalf et al.
Traffic (Copenhagen, Denmark), 9(11), 1905-1914 (2008-09-27)
The neuronal ceroid lipofuscinoses (NCLs, Batten disease) are a group of inherited childhood-onset neurodegenerative disorders characterized by the lysosomal accumulation of undigested material within cells. To understand this dysfunction, we analysed trafficking of the cation-independent mannose 6-phosphate receptor (CI-MPR), which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service