Skip to Content
Merck
All Photos(1)

Key Documents

218739

Sigma-Aldrich

Caspase-1 Substrate X, Fluorogenic

A fluorogenic substrate for detecting caspase-1 (ICE), caspase-4, and caspase-5 activity.

Synonym(s):

Caspase-1 Substrate X, Fluorogenic, Caspase-5 Substrate III, Fluorogenic, Ac-WEHD-AMC

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€251.00

€251.00


Estimated to ship on29 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€251.00

About This Item

Empirical Formula (Hill Notation):
C38H40N8O11
CAS Number:
Molecular Weight:
784.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€251.00


Estimated to ship on29 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

solubility

DMSO: 5 mg/mL

fluorescence

λex 365–380 nm
λem 430–460 nm

shipped in

ambient

storage temp.

−20°C

General description

A fluorogenic substrate for detecting caspase-1 (ICE) (excitation maximum: ~365-380 nm; emission maximum: ~430-460 nm).

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Trp-Glu-His-Asp-AMC

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Thornberry, N.A., et al. 1997. J. Biol. Chem. 272, 17907.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service