Skip to Content
Merck
All Photos(1)

Key Documents

218705

Sigma-Aldrich

Casein Kinase I Inhibitor, D4476

InSolution, ≥95%

Synonym(s):

InSolution Casein Kinase I Inhibitor, D4476

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€205.00

€205.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
1 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C23H18N4O3
Molecular Weight:
398.41
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on05 May 2025Details


Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, potent, and relatively specific ATP-competitive inhibitor of CK1 (IC50 = 200 nM from S. pombe; 0.3 µM for CK1δ) and ALK5 (IC50 = 500 nM). A weak inhibitor of p38α MAP kinase (IC50 = 12 µM) and several other kinases tested, including PKB, SGK, and GSK-3β. Suppresses the site-specific phosphorylation (Ser322 and Ser325) and nuclear exclusion of FOXO1A in H4IIE hepatoma cells. Shown to be ~10-fold more potent than IC261 (Cat. No. 400090; IC50 = 2.5 µM for CK1).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CK1, CK1d
Product competes with ATP.
Reversible: no
Target IC50: 0.2 µM, 0.3 µM against CK1, CK1d, respectively

Packaging

Packaged under inert gas
Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/251 µl) solution of Casein Kinase I Inhibitor, D4476 (Cat. No. 218696) in DMSO

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Other Notes

Rena, G., et al. 2004. EMBO Reports5, 60.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Vural et al.
Journal of cell science, 133(17) (2020-08-02)
Activator of G-protein signaling 3 (AGS3, encoded by GPSM1) was discovered as a one of several receptor-independent activators of G-protein signaling, which are postulated to provide a platform for divergence between canonical and noncanonical G-protein signaling pathways. Similarly, Dishevelled (DVL)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service