Skip to Content
Merck
All Photos(1)

Documents

217277

Millipore

Carboxymethylcellulose, Sodium Salt, Low Viscosity

Substrate for cellulase. Low viscosity (2%, H2O, 25°C): 25 - 75 mPas.

Synonym(s):

Carboxymethylcellulose, Sodium Salt, Low Viscosity

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

form

solid (free-flowing)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to tan

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

General description

Substrate for cellulase. Viscosity: (2%, H2O, 25°C): 25 - 75 mPas.
Substrate for cellulase. Viscosity: 25-75 mPas at 25°C, 2% in H2O.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temprature. Stock solutions are stable for up to 6 months at room temprature.

Other Notes

Christakopoulos, P., et al. 1999. Arch. Biochem. Biophys. 364, 61.
Blanco, A., et al. 1998. Appl. Microbiol. Biotechnol. 50, 48.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service