The product has a solubility of 25 mg/mL in acetonitrile and is also soluble in DMF and DMSO. After reconstitution, the stock can be diluted in a buffer or medium. Please note that it is not a sterile product, so sterile filtration is necessary for stock solutions. It is advisable to store the solution in single-use aliquots for up to 1 week at -20°C, protected from light.
216254
BCECF/AM
A variable mixture of cell-permeable ester derivatives of BCECF that are hydrolyzed by cytosolic esterases to yield intracellularly trapped indicator BCECF.
Synonym(s):
BCECF/AM, 2ʹ,7ʹ- bis-(Carboxyethyl)-5(6ʹ)-carboxyfluorescein Acetoxymethyl Ester, 2ʹ,7ʹ-bis-(Carboxyethyl)-5(6ʹ)-carboxyfluorescein Acetoxymethyl Ester
Select a Size
€199.00
Estimated to ship on25 April 2025Details
Select a Size
About This Item
€199.00
Estimated to ship on25 April 2025Details
Recommended Products
Quality Level
Assay
≥85% (mixture of isomers, HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to yellow
solubility
acetonitrile: 25 mg/mL
DMF: soluble
DMSO: soluble
fluorescence
λex 500-504 nm
λem 525-531 nm
shipped in
ambient
storage temp.
−20°C
SMILES string
O1C3(c4c(cc(c(c4)CCC(=O)OCOC(=O)C)OCOC(=O)C)Oc5c3cc(c(c5)OCOC(=O)C)CCC(=O)OCOC(=O)C)c2c(cc(cc2)C(=O)OCOC(=O)C)C1=O
InChI key
NTECHUXHORNEGZ-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Monitor intracellular pH changes
Packaging
Warning
Reconstitution
Other Notes
Hayashi, H., et al. 1992. Am. J. Physiol. 262, C628.
Sasaki, S., et al. 1992. Biochim. Biophys. Acta 1137, 45.
Miyata, H., et al. 1989. Biochem. Biophys. Res. Commun. 163, 500.
Weiner, I.D., and Hamm, L.L. 1989. Am. J. Physiol.256, F957.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How can product number 216254 be prepared and dissolved?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service