Skip to Content
Merck
All Photos(2)

Documents

215900

Sigma-Aldrich

Carbonic Anhydrase IX/XII Inhibitor

The Carbonic Anhydrase IX/XII Inhibitor controls the biological activity of Carbonic Anhydrase IX/XII. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Carbonic Anhydrase IX/XII Inhibitor, CAIX Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N4O5S
CAS Number:
Molecular Weight:
336.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(S(N)(=O)=O)C=C1)NC2=CC=C(N(=O)=O)C=C2

General description

A cell permeable benzenesulfonamide Carbonic Anhydrase (CA) inhibitor that is selective for human hCA IX and hCA XII (Ki = 0.9 nM and 5.7 nM, respectively) against two other physiologically relevant hCA isoforms (Ki = 23.4 nM and 15 nM for hCA I and hCA II, respectively), as compared with another CA inhibitor AAZ (Ki = 250 nM, 12 nM, 25 nM, and 5.7 nM for hCA I, hCA II, hCA IX, and hCA XII, respectively). It is shown to effectively inhibit the formation of metastases by 4T1 breast cancer cells in a mouse model dose-dependently at pharmacologic concentrations of 15-45 mg/kg.
A cell permeable benzenesulfonamide Carbonic Anhydrase (CA) inhibitor that is selective for human hCA IX and hCA XII (Ki = 0.9 nM and 5.7 nM, respectively) against two other physiologically relevant hCA isoforms (Ki = 23.4 nM and 15 nM for hCA I and hCA II, respectively), as compared with another CA inhibitor AAZ (Ki = 250 nM, 12 nM, 25 nM, and 5.7 nM for hCA I, hCA II, hCA IX, and hCA XII, respectively). It is shown to effectively inhibit the formation of metastases by 4T1 breast cancer cells in a mouse model dose-dependently at pharmacologic concentrations of 15-45 mg/kg.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Pacchiano, F., et al. 2011. J Med Chem.54, 1896.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service