Skip to Content
Merck
All Photos(1)

Key Documents

2140FR

Millipore

Total Protein Extraction Kit

Sign Into View Organizational & Contract Pricing

Select a Size

2140FR
€238.00

€238.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
2140FR
€238.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.32

€238.00


In StockDetails


Request a Bulk Order

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service