Skip to Content
Merck
All Photos(1)

Key Documents

208725

Sigma-Aldrich

Calphostin C, Cladosporium cladosporioides

A cell permeable, highly specific inhibitor of protein kinase C (IC₅₀ = 50 nM) that interacts with the protein′s regulatory domain by competing at the binding site of diacylglycerol and phorbol esters.

Synonym(s):

Calphostin C, Cladosporium cladosporioides, UCN-1028c

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H38O14
CAS Number:
Molecular Weight:
790.76
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown

solubility

DMSO: 1 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C44H38O14/c1-20(56-43(50)22-10-8-7-9-11-22)16-25-31-32-26(17-21(2)57-44(51)58-24-14-12-23(45)13-15-24)42(55-6)40(49)34-28(47)19-30(53-4)36(38(32)34)35-29(52-3)18-27(46)33(37(31)35)39(48)41(25)54-5/h7-15,18-21,45,48-49H,16-17H2,1-6H3

InChI key

LSUTUUOITDQYNO-UHFFFAOYSA-N

General description

A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50 nM) that interacts with the protein’s regulatory domain by competing at the binding site of diacylglycerol and phorbol esters. Does not compete with Ca2+ or phospholipids. At higher concentrations inhibits myosin light chain kinase (IC50 >5 µM), protein kinase A (IC50 >50 µM), protein kinase G (IC50 >25 µM), and p60v-src (IC50 >50 µM). Induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Requires brief exposure to light for activation.
Cell-permeable, highly specific inhibitor of protein kinase C (IC50 = 50 nM) that interacts with the protein′s regulatory domain by competing at the binding site of diacylglycerol and phorbol esters. At higher concentrations inhibits myosin light chain kinase (IC50 >5 µM), protein kinase A (IC50 >50 µM), protein kinase G (IC50 >25 µM), and p60v-src protein tyrosine kinase (IC50 >50 µM). Does not compete with Ca2+ or phospholipids. Also induces apoptotic DNA fragmentation and cell death in HL-60 human promyelocytic leukemia cells. Inhibition of PKC is dependent on exposure to light in cell-free systems. Ordinary fluorescent light is sufficient for full activation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product does not compete with ATP.
Reversible: no
Target IC50: 50 nM against protein kinase C

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Jarvis, W.D., et al. 1994. Cancer Res.54, 1707.
Svetlov, S., and Nigami, S. 1993. Biochim. Biophys. Acta 1177, 75.
Gopalakrishna, R., et al. 1992. FEBS Lett. 314, 149.
Shimamato, H., et al. 1992. Br. J. Pharmacol.107, 282.
Bruns, R.F., et al. 1991. Biochem. Biophys. Res. Commun.176, 288.
Tamaoki, T., et al. 1990. Biotechnology8, 732.
Kobayashi, E., et al. 1989. Biochem. Biophys. Res. Commun.159, 548.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Norman et al.
FEBS letters, 484(3), 179-183 (2000-11-18)
Aggregation by immune complexes of receptors specific for the Fc region of IgG results in their internalisation and disposal by trafficking to lysosomes. We show here that internalisation of FcgammaRI by IFN-gamma treated U937 cells following receptor aggregation by cross-linking
Humberto Muzi-Filho et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 54(6), 1143-1162 (2020-11-18)
Chronic malnutrition (M) affects >1 billion people worldwide. Epidemiological data point to long-term renal and cardiovascular outcomes (e.g. arterial hypertension, cardiorenal syndromes). The renin-angiotensin-aldosterone system (RAAS) has been implicated in the physiopathology of these disturbances, but M-induced alterations in RAAS-modulated
Tomoya Sasahara et al.
iScience, 24(9), 102936-102936 (2021-08-31)
Amyloid β-protein (Aβ) may contribute to worsening of Alzheimer's disease (AD) through vascular dysfunction, but the molecular mechanism involved is unknown. Using ex vivo blood vessels and primary endothelial cells from human brain microvessels, we show that patient-derived Aβ assemblies, termed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service