Skip to Content
Merck
All Photos(2)

Key Documents

205536

Sigma-Aldrich

Cadein1

A cell-permeable isoquinolinium compound that preferentially induces apoptotic cell death in p53-deficient cancer cells with functional MMR (DNA mismatch repair) system.

Synonym(s):

Cadein1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H48F2INO2
Molecular Weight:
655.64
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FC1=CC=CC(F)=C1C[N+](CC2)=C(C3=C2C=C(C(OC)=C3)OC)CCCCCCCCCCCCCCC.[I-]

General description

A cell-permeable isoquinolinium compound that preferentially induces apoptotic cell death in p53-deficient cancer cells with functional MMR (DNA mismatch repair) system, including HCT116-Ch3/E6, HeLa, HT29, and SW620 (viability &lt19 %; 4-6 µM Cadein1 for 24 h), while exhibiting much reduced potency in MMR-/p53+ (RKO and HCT116), MMR-/p53- (DLD-1 and HCT116-E6), and MMR+/p53+ (HCT116-Ch3 and HEK293) cancer cultures (viability = 40 to 92%; 4 µM Cadein1 for 24 h).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Jang, E.R., et al. 2010. J. Biol. Chem.285, 2986.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service