Skip to Content
Merck
All Photos(1)

Key Documents

204878

Sigma-Aldrich

Complement C1r, Human, Activated, Two-Chain Form

Complement C1r, Human, Activated, Two-Chain Form, CAS 80295-34-7, is a native, activated, C1r complement component.

Synonym(s):

Complement C1r, Human, Activated, Two-Chain Form

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€541.00

€541.00


Estimated to ship on25 April 2025Details



Select a Size

Change View
250 μG
€541.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

€541.00


Estimated to ship on25 April 2025Details


Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, albumin, C1s, C4BP, C1-INH, trace

shipped in

wet ice

storage temp.

−70°C

General description

Native, human, activated, C1r complement component. Generally, non-activated form composed of a single-chain glycoprotein. Present in normal human serum at 34 µg/ml. Non-activated C1r is found in circulating blood plasma as a dimer that associates with one C1q molecule and two C1s molecules to form the first component of complement (C1). Ensuing C1q binding to the classical complement pathway activators, each C1r proenzyme monomer is activated by cleavage into two disulfide-linked fragments of M.W. 60,000 and 35,000. The 35,000 peptide contains C1r enzymatic active site. Activated C1r continues activation via the classical pathway by cleaving, and thus activating C1s.

Biochem/physiol Actions

≥80% of C1r will bind with excess C1-INH

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 140 mM NaCl, 10 mM imidazole, 2 mM EDTA, pH 7.4.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Villiers, C.L., et al. 1983. Biochem. J. 215, 369.
Ziccardi, R.J. and Cooper, N.R. 1976. J. Immunol. 116, 496.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service