Skip to Content
Merck
All Photos(1)

Key Documents

203980

Sigma-Aldrich

α-Bungarotoxin

from Bungarus multicinctus, No contaminants by IEF, lyophilized solid, irreversible binder to motor end-plate acetylcholine receptor, Calbiochem®

Synonym(s):

α-Bungarotoxin, Bungarus multicinctus

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€214.00

€214.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C338H529N97O105S11
CAS Number:
Molecular Weight:
7984.12
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€214.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Product Name

α-Bungarotoxin, Bungarus multicinctus, Blocks neuromuscular transmission by irreversible binding to motor end-plate acetylcholine receptor (Kd = 1 pM to 1 nM) but does not depress acetylcholine release from motor nerve endings.

Quality Level

description

Merck USA index - 14, 1488

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

PBS: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

A polypeptide composed of 74 amino acids containing 5 disulfide bridges. Blocks neuromuscular transmission by irreversible binding to the motor end-plate acetylcholine receptor (Kd = 1 nM to 1 pM) but does not depress acetylcholine release from motor nerve endings. Blocks nicotine-induced augmentation in intracellular Ca2+ in PC12 cells (IC50 = 310 nM). Prevents opening of nicotinic receptor-associated ion channels. Reconstitution experiments in Xenopus oocytes have shown the effects of α-Bungarotoxin on neuronal nAChR to be highly specific for the α7-subtype (IC50 = 1.6 nM), but not for the α3β4-subtype (IC50 >3 µM).
Blocks neuromuscular transmission by irreversible binding to motor end-plate acetylcholine receptor (Kd = 1 pM to 1 nM) but does not depress acetylcholine release from motor nerve endings. Blocks nicotine-induced increase of intracellular Ca2+ in PC12 cells (IC50 = 310 nM), and prevents opening of nicotinic receptor-associated ion channels. Reconstitution experiments in Xenopus oocytes have shown the effects of α-bungarotoxin on neuronal nAChR to be highly specific for the α7-subtype (IC50 = 1.6 nM), but not for the α3β4-subtype (IC50 >3 µM).

Biochem/physiol Actions

Cell permeable: no
Kd = 1 pM to 1 nM for motor end-plate acetylcholine receptor
Primary Target
Motor end-plate acetylcholine receptor
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Sequence

H-Ile-Val-Cys³-His-Thr-Thr-Ala-Thr-Ser-Pro-Ile-Ser-Ala-Val-Thr-Cys¹⁶-Pro-Pro-Gly-Glu-Asn-Leu-Cys²³-Tyr-Arg-Lys-Met-Trp-Cys²⁹-Asp-Ala-Phe-Cys³³-Ser-Ser-Arg-Gly-Lys-Val-Val-Glu-Leu-Gly-Cys⁴⁴-Ala-Ala-Thr-Cys⁴⁸-Pro-Ser-Lys-Lys-Pro-Tyr-Glu-Glu-Val-Thr-Cys⁵⁹-Cys⁶⁰-Ser-Thr-Asp-Lys-Cys⁶⁵-Asn-Pro-His-Pro-Lys-Gln-Arg-Pro-Gly-OH (disulfide bonds: 3 → 23; 16 → 44; 29 → 33; 48 → 59; 60 → 65)

Physical form

Supplied as an acetate salt

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Analysis Note

Contaminants: None detectable by IEF

Other Notes

Lopez, M.G., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 14184.
Zhang, Z.W., et al. 1994. Neuron 12, 167.
Bambrick, L.L., and Gordon, T. 1992. J. Physiol.449, 479.
Lin, S.R., and Chang, C.C. 1992. Biochim. Biophys. Acta1159, 255.
Motomura, M., et al. 1992. Neurosci. Lett.143, 139.
Sorenson, E.M., and Chiappinelli, V.A. 1992. J. Comp. Neurol.323, 1.
Ruan, K.H., et al. 1990. Proc. Natl. Acad. Sci. USA87, 6156.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service