Skip to Content
Merck
All Photos(1)

Key Documents

203389

Sigma-Aldrich

Blebbistatin, Racemic

≥97% (HPLC), liquid, Myosin II inhibitor, Calbiochem®

Synonym(s):

InSolutionBlebbistatin, Racemic

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€214.00

€214.00


Estimated to ship on16 April 2025Details



Select a Size

Change View
5 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C18H16N2O2
Molecular Weight:
292.33
UNSPSC Code:
12352200
NACRES:
NA.77

€214.00


Estimated to ship on16 April 2025Details


Product Name

Blebbistatin, Racemic, InSolution, ≥97%, 50 mM in 90% DMSO, reversible inhibitor of nonmuscle myosin II

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable compound that acts as a selective, potent, and reversible inhibitor of non-muscle myosin II. Inhibits the ATPase and gliding motility of human platelets (≤100 µM) without affecting myosin light chain kinase (MLCK) activity. Has been shown to block cell blebbing and to rapidly disrupt directed cell migration and cytokinesis in vertebrate cells. Does not disrupt mitosis or affect contractile ring assembly.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ATPase
Product does not compete with ATP.
Reversible: yes
Target IC50: <100 µM inhibiting the ATPase and gliding motility of human platelets

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Physical form

A 50 mM (5 mg/342 µl) solution of (±)-Blebbistatin (Cat. No. 203390) in 90% DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Shu, S., et al. 2005. Proc. Natl. Acad. Sci. USA102, 1472.
Kovacs, M., et al. 2004. J. Biol. Chem.279, 35557.
Straight, A.F., et al. 2003. Science299, 1743.
Cheung, A., et al. 2001. Mol. Biol. Cell Suppl.12, 271a.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin S Weiß et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 11(19), e2305947-e2305947 (2024-03-13)
Tissue homeostasis and disease states rely on the formation of new blood vessels through angiogenic sprouting, which is tightly regulated by the properties of the surrounding extracellular matrix. While physical cues, such as matrix stiffness or degradability, have evolved as
Kevin X Lau et al.
Nature communications, 11(1), 2420-2420 (2020-05-18)
Archetypal human pluripotent stem cells (hPSC) are widely considered to be equivalent in developmental status to mouse epiblast stem cells, which correspond to pluripotent cells at a late post-implantation stage of embryogenesis. Heterogeneity within hPSC cultures complicates this interspecies comparison.
Cell segregation via differential collision modes between heterotypic cell populations.
Lucia, et al.
Molecular Biology of the Cell, 33, ar129-ar129 (2023)
Catarina Domingues et al.
Frontiers in cell and developmental biology, 8, 678-678 (2020-09-10)
The mechanical properties of the extracellular environment are interrogated by cells and integrated through mechanotransduction. Many cellular processes depend on actomyosin-dependent contractility, which is influenced by the microenvironment's stiffness. Here, we explored the influence of substrate stiffness on the proteome
Lawrence J Dooling et al.
Nature biomedical engineering, 7(9), 1081-1096 (2023-04-25)
In solid tumours, the abundance of macrophages is typically associated with a poor prognosis. However, macrophage clusters in tumour-cell nests have been associated with survival in some tumour types. Here, by using tumour organoids comprising macrophages and cancer cells opsonized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service