Skip to Content
Merck
All Photos(1)

Key Documents

20-210

Millipore

Catch and Release Wash Buffer, 10X

Sign Into View Organizational & Contract Pricing

Select a Size

15 ML
€90.50

€90.50


Estimated to ship on21 March 2025Details



Select a Size

Change View
15 ML
€90.50

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NB.22

€90.50


Estimated to ship on21 March 2025Details


form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

shipped in

wet ice

Application

For use with the Catch and Release reversible immunoprecipitation system for cell lysis and dilution, and to wash Catch and Release Spin Columns during the immunoprecipitation protocol.
Research Category
All

Physical form

Buffer, pH 7.4, containing the following detergents: 10% NP-40 and 2.5% deoxycholic acid

Storage and Stability

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service