Skip to Content
Merck
All Photos(1)

Key Documents

20-135

Millipore

Autorad Orientation Markers

Western blotting, dot/slot blotting

Sign Into View Organizational & Contract Pricing

Select a Size

5 UNITS
€97.80

€97.80


Estimated to ship on29 April 2025Details



Select a Size

Change View
5 UNITS
€97.80

About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

€97.80


Estimated to ship on29 April 2025Details


Quality Level

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

detection method

chemiluminescent

shipped in

ambient

Physical form

One packet containing 5 markers

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service