Skip to Content
Merck
All Photos(1)

Key Documents

20-133-M

Sigma-Aldrich

PKC Lipid Activator

Synonym(s):

PKC activator, Protein kinase C activator

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€204.00
5 ML
€235.00

€204.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€204.00
5 ML
€235.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

€204.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PRKCA(5578)

General description

Protein kinase C (PKC) belongs to a family of serine/ threonine kinases.[1] It comprises a regulatory domain at the N-terminus and a catalytic domain at the C-terminus. The regions are divided into conserved regions (C1–C4) and regions that differ between isoforms (V1–V5). PKC has a common pseudosubstrate domain in the regulatory region.[1]

Application

For use in kinase assays.
Research Category
All

Biochem/physiol Actions

Protein kinase C (PKC) is activated by diacylglycerol (DAG) and acts as an intracellular receptor of tumor-promoting phorbol esters. It regulates several cell functions and signal transduction pathways through cell migration and polarity, differentiation, proliferation, and cell death. PKC is part of a large group of phospholipid-dependent enzymes, which catalyze the covalent transfer of phosphate from ATP to serine and threonine residues of proteins.[1]

Physical form

20mM MOPS, pH 7.2, 25mM β-glycerolphosphate, 1mM sodium orthovanadate, 1mM dithiothreitol, 1mM CaCl2.

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Breitkreutz et al.
Journal of cancer research and clinical oncology, 133(11), 793-808 (2007-07-31)
The protein kinase C (PKC) family represents a large group of phospholipid dependent enzymes catalyzing the covalent transfer of phosphate from ATP to serine and threonine residues of proteins. Phosphorylation of the substrate proteins induces a conformational change resulting in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service