Skip to Content
Merck
All Photos(1)

Key Documents

1.99004

Supelco

Buffer solution

(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (25°C) Certipur®

Synonym(s):

Boric acid/potassium chloride/sodium hydroxide buffer solution, pH 10.0 calibration buffer

Sign Into View Organizational & Contract Pricing

Select a Size

30 X 30 ML
€112.00

€112.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
30 X 30 ML
€112.00

About This Item

UNSPSC Code:
12161705
NACRES:
NA.25

€112.00


Available to ship on03 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

pH

10.0 (25 °C in H2O)

bp

100 °C/1013 hPa

density

1.0044 g/cm3 at 25 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The boric acid/potassium chloride/sodium hydroxide certified buffer solution is gravimetrically prepared using high-quality reagents and water at a registered calibration laboratory. Its pH value of 10.00 at 25°C is measured using a combined glass electrode following the 5-point calibration method. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. The pH of the buffer is strongly dependent on the temperature and therefore a constant temperature must be maintained during its use.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used as a standard to calibrate pH measuring instruments or pH electrodes and as a control to measure the pH value of other solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
  • Ready-to-use solution available in 30 mL sachets
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide 35
  • Accurate up to +/- 0.03 at 25°C

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service