Skip to Content
Merck
All Photos(1)

Key Documents

1.94959

Supelco

Ammonium dichromate

EMPLURA®

Synonym(s):

Ammonium dichromate, Ammonium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Cr2O7
CAS Number:
Molecular Weight:
252.06
MDL number:
UNSPSC Code:
12352302
Pricing and availability is not currently available.

vapor density

8.7 (vs air)

product line

EMPLURA®

mp

170 °C (dec.) (lit.)

storage temp.

room temp

SMILES string

N.N.O[Cr](=O)(=O)O[Cr](O)(=O)=O

InChI

1S/2Cr.2H3N.2H2O.5O/h;;2*1H3;2*1H2;;;;;/q2*+1;;;;;;;;;/p-2

InChI key

XAQCJVGGJJFLPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service