Skip to Content
Merck
All Photos(1)

Documents

1.94608

Supelco

Isoamyl alcohol

for analysis EMPARTA® ACS

Synonym(s):

3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
MDL number:
UNSPSC Code:
12352001

grade

ACS reagent

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

autoignition temp.

644 °F

expl. lim.

1.2-9 %, 100 °F

refractive index

n20/D 1.406 (lit.)

bp

130 °C (lit.)

mp

−117 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service