Skip to Content
Merck
All Photos(1)

Key Documents

1.94607

Supelco

1,2-Dichloroethane

for analysis EMPARTA® ACS

Synonym(s):

1,2-Dichloroethane, Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
MDL number:
UNSPSC Code:
12352101
grade:
ACS reagent
bp:
83 °C (lit.)
vapor pressure:
87 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

autoignition temp.

775 °F

expl. lim.

15.6 %

dilution

(for routine analytical applications)

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service