Skip to Content
Merck
All Photos(1)

Key Documents

1.93242

Supelco

Mercury(II) nitrate monohydrate

for analysis EMPARTA® ACS

Synonym(s):

Mercury(II) nitrate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(NO3)2 · H2O
CAS Number:
Molecular Weight:
324.60 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
grade:
ACS reagent

grade

ACS reagent

storage temp.

room temp

SMILES string

[Hg+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O

InChI

1S/Hg.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

KVICROHOONHSRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93242.0121: 100 g, Minimum order quantity: 20

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service