Skip to Content
Merck
All Photos(1)

Key Documents

189738

Sigma-Aldrich

Streptavidin, TEXAS RED® Conjugate

This Streptavidin, TEXAS RED® Conjugate is validated for use in ELISA, Flow Cytometry, Immunohistochemistry.

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€262.00

€262.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€262.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.47

€262.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze
protect from light

shipped in

ambient

storage temp.

2-8°C

General description

Streptavidin conjugated to TEXAS RED® fluorescent dye. Specific for biotin, biotinylated proteins, or biotinylated antibodies.

Application

ELISA (1:2000-1:5000)

Flow Cytometry (1:50-1:200)

Immunohistochemistry (1:50-1:200)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM bicarbonate-borate buffer, 0.9% NaCl, 5 mg/ml BSA, pH 8.1.

Reconstitution

Do not dilute the entire sample at one time. Withdraw aliquots as needed with a micropipette and dilute with 1% solution of BSA in 50 mM sodium phosphate buffer, pH 7.4 and 0.05% sodium azide.

Other Notes

Biotin activity, absorbance ratio, and label:protein molar ratio are determined on a lot-specific basis. Use 1 to 3 µg of the conjugate per 1 x 106 cells suspended in a volume of 0.1 ml. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Texas Red is a registered trademark of Life Technologies

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service