Skip to Content
Merck
All Photos(1)

Key Documents

182755

Millipore

ASB-16

A zwitterionic amidosulfobetaine detergent useful for solubilizing proteins visualized by 2D-electrophoresis.

Synonym(s):

ASB-16

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€833.00

€833.00


Estimated to ship on26 March 2026Details


Request a Bulk Order

Select a Size

Change View
25 G
€833.00

About This Item

Empirical Formula (Hill Notation):
C24H50N2O4S
CAS Number:
Molecular Weight:
462.73
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

€833.00


Estimated to ship on26 March 2026Details


Request a Bulk Order

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

20% ethanol: 1 mg/mL
water: sparingly soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+](CCCNC(=O)CCCCCCCCCCCCCCC)(C)C

InChI

1S/C24H50N2O4S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-19-24(27)25-20-17-21-26(2,3)22-18-23-31(28,29)30/h4-23H2,1-3H3,(H-,25,27,28,29,30)

InChI key

CLCSYZQBLQDRQU-UHFFFAOYSA-N

General description

A zwitterionic amidosulfobetaine detergent useful for solubilizing proteins visualized by 2D-electrophoresis. The increased solubilization produced by this detergent enables the identification of previously undetected membrane proteins.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Other Notes

Flatman, P. W., et al. 2005. J. Physiol.563, 421.
Herbert, B. 1999. Electrophoresis20, 660.
Chevallet, M., et al. 1998. Electrophoresis19, 1901.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service