Skip to Content
Merck
All Photos(1)

Key Documents

178455

Sigma-Aldrich

Apolipoprotein A-II, Human Plasma, High-Density Lipoprotein

Native apolipoprotein A-II from human plasma. The second most abundant protein of HDL. Binds to phospholipids during lipoprotein metabolism. Displaces LCAT bound to lipoprotein.

Synonym(s):

Apo A-II

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€401.00

€401.00


Please contact Customer Service for Availability


Select a Size

Change View
500 μG
€401.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€401.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

Native apolipoprotein A-II from human plasma. Shown to bind to phospholipids during lipoprotein metabolism. Displaces lecithin-cholesterol acyltransferase bound to lipoprotein. Influences HDL functional states and contributes to arteriosclerosis.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Store undiluted.

Other Notes

Fu, L., et al. 2001. J. Histochem. Cytochem.49, 739.
Hedrick, C.C., and Lusis, A.J. 1994. Can. J. Cardiol.10, 453.
Breslow, J.L. 1993. Proc. Natl. Acad. Sci. USA 90, 8314.
Schultz, J.R., et al. 1992. J. Biol. Chem.267, 21630.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrzej S Pitek et al.
PloS one, 7(7), e40685-e40685 (2012-07-26)
It is now well established that the surface of nanoparticles (NPs) in a biological environment is immediately modified by the adsorption of biomolecules with the formation of a protein corona and it is also accepted that the protein corona, rather

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service