Skip to Content
Merck
All Photos(1)

Key Documents

178278

Sigma-Aldrich

Apigenin

Induces the reversion of transformed phenotypes of v-H-ras-transformed NIH 3T3 cells at low concentration (12.5 µM).

Synonym(s):

Apigenin, 4ʹ,5,7-Trihydroxyflavone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€53.20

€53.20


Estimated to ship on08 May 2025Details



Select a Size

Change View
5 MG
€53.20

About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
MDL number:
UNSPSC Code:
12352212
NACRES:
NA.77

€53.20


Estimated to ship on08 May 2025Details


Quality Level

description

Merck USA index - 14, 730

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

solubility

hot ethanol: 1 mg/mL
DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[o]1c2c([c](cc1c3ccc(cc3)O)=O)c(cc(c2)O)O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H

InChI key

KZNIFHPLKGYRTM-UHFFFAOYSA-N

General description

A plant flavonoid that reverses the transformed phenotypes of various ras-transformed cells, apparently due to inhibition of mitogen activated protein (MAP) kinase-associated signal transduction pathways. Treatment of v-H-ras-transformed NIH 3T3 cells with apigenin (12.5 µM) has been shown to induce dephosphorylation of p44mapk and reduction in MAP kinase activity. Has also been reported to inhibit the proliferation of malignant tumor cells by G2/M arrest and to induce morphological differentiation.
Induces the reversion of transformed phenotypes of v-H-ras-transformed NIH 3T3 cells at low concentration (12.5 µM). This finding has been attributed to the inhibition of MAP kinase activity. Inhibits the proliferation of malignant tumor cells by G2/M arrest and induces morphological differentiation.

Biochem/physiol Actions

Cell permeable: no
Primary Target
MAP kinase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 4 months at -20°C.

Other Notes

Kuo, M.L., and Yang, N.C. 1995. Biochem. Biophys. Res. Commun.212, 767.
Chaumontet, C., et al. 1994. Carcinogenesis 15, 2325.
Kuo, M.L., et al. 1994. Cancer Lett.87, 91.
Sato, F., et al. 1994. Biochem. Biophys. Res. Commun.204, 578.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service