Skip to Content
Merck
All Photos(1)

Key Documents

178276

Sigma-Aldrich

Apicidin, Fusarium sp.

A potent, cell-permeable inhibitor of histone deacetylase (IC₅₀= 700 pM for parasitic histone deactetylase) that also exibits antiprotozoal and potential anti-malarial properties.

Synonym(s):

Apicidin, Fusarium sp., cyclo-[L-(2-Amino-8-oxodecanoyl)-L-(N-methoxytryptophan)-L-isoleucyl-D-pipecolinyl], HDAC Inhibitor XI

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€194.00
5 MG
€482.00

€194.00


Estimated to ship on02 May 2025Details



Select a Size

Change View
1 MG
€194.00
5 MG
€482.00

About This Item

Empirical Formula (Hill Notation):
C34H49N5O6
Molecular Weight:
623.78
UNSPSC Code:
12352200
NACRES:
NA.77

€194.00


Estimated to ship on02 May 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Highly Toxic (H)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service