Skip to Content
Merck
All Photos(1)

Key Documents

17-610

Sigma-Aldrich

Magna ChIP® A - Chromatin Immunoprecipitation Kit

Sign Into View Organizational & Contract Pricing

Select a Size

22 ASSAYS
€649.00

€649.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
22 ASSAYS
€649.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

€649.00


Please contact Customer Service for Availability

Request a Bulk Order

packaging

kit of 22 assay(s)

Quality Level

manufacturer/tradename

Magna ChIP®

technique(s)

immunoprecipitation (IP): suitable

General description

Chromatin Immunoprecipitation (ChIP) is an important technique allowing the researcher to analyze in vivo interactions of proteins with genomic DNA. Any chromatin-associated or DNA binding protein can be analyzed with this technique, provided a good antibody to the protein exists. One can measure different proteins localized to a specific region of the genome, or the genome wide distribution of a specific protein. Another powerful application of this technique is to analyze changes in histone modifications that correlate with processes like transcription, mitosis or DNA repair.

Application

Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads.

Features and Benefits

  • Faster:Magnetic protein A beads allow for the entire ChIP protocol to be done in as little as a day! All reagents to process your samples are included - you don′t have to spend valuable time making them.
  • Easier: Spin columns make DNA purification easier and more reliable - no more messy phenol-chloroform extractions.

Packaging

Two boxes containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions. Supplied buffers are sufficient to generate chromatin from up to five 15 cm plates of cultured cells, each plate providing up to 10 chromatin preparations (varies with cell and assay type).

Components

  • Magnetic Protein A Beads
  • ChIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • Cell Lysis Buffer
  • Nuclear Lysis Buffer
  • ChIP Elution Buffer (w/o Proteinase K)
  • 10X Glycine
  • 10X PBS
  • Protease Inhibitor Cocktail II
  • Proteinase K
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 1 year from date of shipment when stored as directed.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service