172104
Anaphase-Promoting Complex Inhibitor, TAME
The Anaphase-Promoting Complex Inhibitor, TAME, also referenced under CAS 1784-03-8, controls the biological activity of Anaphase-Promoting Complex. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Synonym(s):
Anaphase-Promoting Complex Inhibitor, TAME, TAME hydrochloride, APC/C Inhibitor I, APC/C-Cdc20 Interaction Inhibitor I, Cyclosome Inhibitor I
Select a Size
€74.00
Estimated to ship on11 April 2025Details
Select a Size
About This Item
€74.00
Estimated to ship on11 April 2025Details
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
[S](=O)(=O)(NC(CCC[N+H]=C(N)N)C(=O)OC)c1ccc(cc1)C.[Cl-]
InChI
1S/C14H22N4O4S.ClH/c1-10-5-7-11(8-6-10)23(20,21)18-12(13(19)22-2)4-3-9-17-14(15)16;/h5-8,12,18H,3-4,9H2,1-2H3,(H4,15,16,17);1H
InChI key
JIQFFACVQXXHMY-UHFFFAOYSA-N
General description
Packaging
Warning
Reconstitution
Other Notes
Zeng, X., et al. 2010. Cancer Cell18, 382.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service