Skip to Content
Merck
All Photos(1)

Key Documents

171375

Millipore

Amphotericin B, Streptomyces sp.

A polyene antifungal antibiotic that non-specifically induces loss of low molecular weight substances from cells. Binds to sterols and disrupts osmotic integrity of fungal membrane. Causes suppression of bone marrow progenitor cells.

Synonym(s):

Amphotericin B, Streptomyces sp., Fungizone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

>86% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 30-40 mg/mL
DMF: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5-,9-7-,10-8-,13-11-,14-12-,17-15-,18-16-/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-ZNVUZQDLSA-N

General description

A polyene antifungal antibiotic that non-specifically induces loss of low molecular weight substances from cells. Binds to sterols and disrupts osmotic integrity of fungal membrane. Causes suppression of bone marrow progenitor cells.
An antifungal agent. A polyene antibiotic that nonspecifically induces the loss of low molecular weight substances from cells. Causes suppression of bone marrow progenitor cells. Probably forms channels by complexing with membrane cholesterol. Effective against fungi and yeast at concentrations of 1-10 µg/ml.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C. Solutions in tissue culture media are stable for 3 days at 37°C.

Other Notes

Tollemar, J., et al. 1995. Transplantation 59, 45.
Charak, B.S., et al. 1994. Br. J. Haematol. 88, 693.
Barth, C., et al. 1993. J. Biophys.64, 92.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service