1.70320
Germanium ICP standard
traceable to SRM from NIST (NH₄)₂GeF₆ in H₂O 1000 mg/l Ge Certipur®
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
product line
Certipur®
concentration
1000 mg/L Ge in H2O
technique(s)
ICP: suitable
pH
0.5 (20 °C in H2O)
density
1.000 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
single component solution
storage temp.
15-25°C
Related Categories
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Application
- Environmental Monitoring of Heavy Metals: Germanium ICP standards play a crucial role in the preliminary screening of soils for natural radioactivity and metal(loid) content, particularly around decommissioned industrial sites. This application highlights the importance of high-purity germanium standards in environmental health research to ensure community safety and regulatory compliance (Huang et al., 2022).
- Improvement of Iodine Analysis Methods: The reliability of AOAC Official Method 2012.15 for iodine, which often employs germanium as an internal standard in ICP-MS analyses, underscores the standard′s role in enhancing methodological accuracy in biopharmaceutical applications. This improvement is crucial for ensuring the quality and safety of pharmaceutical products (Hieda et al., 2022).
- Advancements in Photochemical Vapor Generation: Research on the synergistic effects from cobalt/chloride ions and air-liquid interfaces in the generation of germanium hydrides showcases the germanium ICP standard′s application in developing novel analytical techniques. This method provides a more efficient and sensitive approach to trace metal analysis in complex matrices (Yu et al., 2022).
- Serum Lithium Quantification: Germanium ICP standards are pivotal in the standard addition method for quantifying serum lithium via inductively coupled plasma mass spectrometry. This technique is essential in clinical biochemistry for monitoring therapeutic levels of lithium, demonstrating the standard′s role in clinical diagnostics (Fan et al., 2022).
- Environmental Water Analysis: The speciation of germanium in environmental water using hydride generation and ICP-MS/MS indicates the utility of germanium ICP standards in environmental science. This method helps in the accurate and sensitive determination of germanium levels, vital for assessing pollution and its ecological impacts (Garcia-Figueroa et al., 2021).
Analysis Note
Other Notes
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service