Skip to Content
Merck
All Photos(1)

Documents

164545

Sigma-Aldrich

7-Amino-4-methylcoumarin

≥98% (HPLC), solid

Synonym(s):

7-Amino-4-methylcoumarin, AMC, Coumarin 120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

product name

7-Amino-4-methylcoumarin, Reagent used to prepare fluorogenic 7-amido-4-methylcoumarin (AMC) based substrates for the detection of proteolytic enzyme activity.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
acetone: 10 mg/mL
DMF: soluble

fluorescence

λex 365-380 nm
λem 430-460 nm

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C10H9NO2/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5H,11H2,1H3

InChI key

GLNDAGDHSLMOKX-UHFFFAOYSA-N

General description

Reagent used to prepare fluorogenic 7-amido-4-methylcoumarin (AMC) based substrates for the detection of proteolytic enzyme activity. Useful as a reference standard in enzyme assays.
Reagent used to prepare fluorogenic 7-amino-4-methylcoumarin (AMC) based substrates for the detection of proteolytic enzymes (excitation maximum: ~365-380 nm; emission maximum: ~430-460 nm). Also useful as a reference standard in enzyme assays.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Detection of proteolytic enzyme activity
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMF and DMSO stock solutions are stable for up to 6 months at -20°C. Acetone stock solutions are stable for up to 1 week at 4°C or -20°C.

Other Notes

Kanaoka, Y., et al. 1982. Chem. Pharm. Bull. 30, 1485.
Zimmerman, M., et al. 1977. Anal. Biochem. 78, 47.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service