Skip to Content
Merck
All Photos(1)

Key Documents

1.50820

Supelco

LiChrospher® Si 60 (5 µm) HPLC Columns

L x I.D. 125 mm x 4 mm LiChroCART HPLC cartridge, manuCART holder required for use

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€581.00

€581.00


Estimated to ship on17 April 2025Details



Select a Size

Change View
1 UNIT
€581.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

€581.00


Estimated to ship on17 April 2025Details


Product Name

LiChrospher® Si 60 (5 µm) LiChroCART 125-4, suitable for HPLC

material

stainless steel column

Quality Level

Agency

suitable for USP L3

product line

LiChrospher®

feature

endcapped: no

parameter

250 bar pressure
60 °C max. temp.

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable (Low - Medium)

column L × I.D.

12.5 cm × 4 mm

surface area

350 m2/g

matrix

fully porous particle

matrix active group

Silica bonding phase

particle size

5 μm

pore size

60 Å pore size

operating pH range

2-7.5

separation technique

hydrophilic interaction (HILIC)
normal phase

storage temp.

no temp limit

General description

HPLC cartridge
LiChrospher® Si 60 is a versatile HPLC spherical silica sorbent with a pore diameter of 60A and a particle size of 5µm for normal phase chromatography: This sorbent is packed into a LiChroCART 125-4 HPLC cartridge. The connection to the HPLC system is being accomplished by use of the manu-CART NT cartrige holder (1.51486).

Application

  • Recent Analytical Applications of LiChrospher® RP-8 (5µm) LICHROCART 150-4.6: Due to its unique properties, the LiChrospher® RP-8 (5µm) LICHROCART 150-4.6 is widely used in various analytical chemistry applications, including HPLC separation of complex mixtures, pharmaceutical compound analysis, and environmental monitoring. The column provides excellent resolution and peak symmetry, making it ideal for both qualitative and quantitative analyses. Researchers have utilized this column for the separation of peptides, proteins, and other biomolecules, contributing to advancements in biochemistry and molecular biology.

Analysis Note

Pressure: ≤ 40 bar
Theoretical Plates (N/m) (2-Nitrotoluene): ≥ 50000
Symmetry (2-Nitrotoluene): 0.8 - 1.4
Capacity factor (2-Nitrotoluene): ≤ 4.0

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service