Skip to Content
Merck
All Photos(1)

Documents

1.50227

Supelco

MANU-CART® 30 mm HPLC cartridge holder

for LiChroCART® 30-2, 30-3 and 30-4 mm HPLC-cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

manu-CART® 30 mm cartridge holder for LiChroCART® 30-2, 30-3 and 30-4 mm HPLC-cartridges,

technique(s)

HPLC: suitable

Quality Level

storage temp.

no temp limit

Related Categories

General description

manu-CART holder 30 mm for 30-2, 30-3 and 30-4 LiChroCART cartridges.

Analysis Note

visual control: passes test
requirement of the german Zusatzstoff-Verkehrsverordnung.

Legal Information

LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service