Skip to Content
Merck
All Photos(1)

Documents

1490-OP

Millipore

Acrylamide/Bis-acrylamide

OmniPur® Grade, 40% solution, 37.5:1 ratio, Suitable for protein separations

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

color

clear colorless

shipped in

wet ice

General description

Suitable for protein seperations. Molecular biology grade. Reconstitute with deionized water to desired final percentage. Premixed solutions offer standard ratios of acrylamide to bis-acrylamide for convenience and safety. The hazard of handling powdered acrylamide is eliminated with the solutions.

Specifications:
DNase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service