Skip to Content
Merck
All Photos(2)

Key Documents

14-803-M

Sigma-Aldrich

Insulin Receptor, activated, 10 µg

Autoactivated on column by incubating with Mg/ATP, excess ATP & MgAc removed by multiple column wash steps.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€492.00
250 μG
€3,770.00

€492.00


Estimated to ship on24 April 2025Details



Select a Size

Change View
10 μG
€492.00
250 μG
€3,770.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€492.00


Estimated to ship on24 April 2025Details


biological source

human

Quality Level

100
300

mol wt

Mw 38.6 kDa

manufacturer/tradename

Upstate®

NCBI accession no.

General description

N-terminal 6His-tagged, recombinant, human Insulin Receptor amino acids 1005-1310, expressed by baculovirus in Sf21 insect cells. Autoactivated on column by incubating with Mg/ATP, excess ATP and MgAc removed by multiple column wash steps.
Product Source: Expressed by baculovirus in Sf21 insect cells.

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: IR
Target Sub-Family: TK

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-803M when ordering the 250μg size.

Physical form

50mM Tris/HCl pH7.5, 150mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol, 5mM β-glycerophosphate, 1mM Na3VO4. Frozen solution.
Ni2+/NTA-agarose

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service