Skip to Content
Merck
All Photos(1)

Key Documents

14-801-M

Sigma-Aldrich

Tec Protein, activated, 10 µg

Autoactivated by incubating with Mg/ATP, excess ATP & MgAc removed by gel filtration.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€577.00
250 μG
€3,470.00

€577.00


Estimated to ship on03 April 2025Details



Select a Size

Change View
10 μG
€577.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€577.00


Estimated to ship on03 April 2025Details


biological source

human

Quality Level

100
300

mol wt

Mw 57 kDa

manufacturer/tradename

Upstate®

NCBI accession no.

UniProt accession no.

Gene Information

human ... TEC(100124696)

General description

N-terminal 6His-tagged, recombinant, human Tec amino acids 174-end. Autoactivated by incubating with Mg/ATP, excess ATP and MgAc removed by gel filtration.

Biochem/physiol Actions

Protein Target: Tec

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-801M when ordering the 250μg size.

Physical form

50 mM Tris/HCl pH7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 5 mM β-glycerophosphate, 1 mM Na3VO4. Frozen solution.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service