Skip to Content
Merck
All Photos(1)

Key Documents

14-794-M

Sigma-Aldrich

PI 3-Kinase (p110β/p85α) murine, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€543.00
250 μG
€3,470.00

€543.00


Estimated to ship on10 April 2025Details



Select a Size

Change View
10 μG
€543.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.32

€543.00


Estimated to ship on10 April 2025Details


biological source

mouse

Quality Level

mol wt

Mw 126 kDa (p110β MW)
Mw 84 kDa (p85α MW)

manufacturer/tradename

Upstate®

NCBI accession no.

UniProt accession no.

General description

Co-expressed by baculovirus in Sf21 insect cells.
Complex of N-terminal 6His-tagged, recombinant murine p110β, full length and untagged, recombinant, murine p85α, full length.

Application

Complex of N-terminal 6His-tagged, recombinant murine p110β, full length & untagged, recombinant, murine p85α, full length.

Biochem/physiol Actions

  • Protein Target: PI3Kβ/p85α
  • Target Sub-Family:PI3K/PI4K

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-794M when ordering the 250μg size.

Physical form

50 mM Tris/HCl pH7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 0.2 mM PMSF, 1 mM benzamidine, 0.1% 2-mercaptoethanol. Frozen solution.

Format: Ni2+/NTA-agarose

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service