Skip to Content
Merck
All Photos(1)

Key Documents

14-788-M

Sigma-Aldrich

PI3 Kinase (p110β/p85β) Murine, 250 µg

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€3,470.00

€3,470.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€3,470.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target PI3Kβ/p85β

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

mouse ... PIK3CB(74769)

General description

FUNCTION: SwissProt: P42338 # Phosphorylates PtdIns, PtdIns4P and PtdIns(4,5)P2 with a preference for PtdIns(4,5)P2.
SIZE: 1070 amino acids; 122762 Da
SUBUNIT: Heterodimer of a p110 (catalytic) and a p85 (regulatory) subunit.
TISSUE SPECIFICITY: Expressed ubiquitously.
SIMILARITY: SwissProt: P42338 ## Belongs to the PI3/PI4-kinase family. & Contains 1 PI3K/PI4K domain.

Packaging

Also available in 10 μg size (cat#14-788) and in bulk (cat# 14-788-K).

Physical properties

p110β MW = 125.5kDa, p85β MW = 81.4kDa

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service