Skip to Content
Merck
All Photos(1)

Key Documents

14-749-M

Sigma-Aldrich

TAO1 Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, human TAO1 amino acids 1-327. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€436.00
250 μG
€3,470.00

€436.00


Estimated to ship on23 April 2025Details



Select a Size

Change View
10 μG
€436.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€436.00


Estimated to ship on23 April 2025Details


biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

Assay

93% (Coomassie blue staining, SDS-PAGE)

specific activity

1179 U/mg

mol wt

Mw 41 kDa

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... TAOK1(57551)

General description

N-terminal 6His-tagged, recombinant, human TAO1 amino acids 1-327.
Product Source: Expressed by baculovirus in Sf21 cells.

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: TAO1
Target Sub-Family: STE

Quality

Routinely evaluated by phosphorylation of 0.8 mg/mL myelin basic protein. Purity was assessed by SDS-PAGE and Coomassie blue staining using 3 μg of TAO1.

Unit Definition

Specific Activity: where one unit of TAO1 activity is defined as 1 nmol phosphate incorporated into 0.8 mg/mL myelin basic protein per minute at 30°C with a final ATP concentration of 100 μM.

Physical form

10 μg of enzyme in 100 μL of 50 mM Tris/HCl pH7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij 35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Ni2+/NTA-agarose

Storage and Stability

Stable for 6 months at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hutchison et al.
The Journal of biological chemistry, 273(44), 28625-28632 (1998-10-24)
Several components of the budding yeast pheromone-response pathway are conserved in mammalian mitogen-activated protein (MAP) kinase pathways. Thus, we used degenerate oligonucleotides derived from the sequence of the Saccharomyces cerevisiae protein kinase Ste20p to amplify related sequences from the rat.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service