closure type Red screw cap with 3 loci ready-to-use
packaging
pack of 6 bottles
technique(s)
direct inoculation: suitable microbiological culture: suitable
bottle capacity
500 mL
bottle filling volume
500 mL
capacity
500 mL
pH
6.9-7.3
application(s)
pharmaceutical sterility testing
storage temp.
2-25°C
suitability
nonselective for bacteria
General description
Fluid D is a washing solution for sterility testing of pharmaceutical samples containing lecithin or oil using the membrane filtration method. Rinse the membrane with Fluid D after filtration and provide a suitable medium to observe microbial growth.
Application
Fluid D is suitable for rinsing the solutions containing oil or lecithin during sterility testing by membrane filtration. It is also suitable for the removal of antimicrobial activity of non-sterile products by membrane filtration. The fluid is also excellent for rinsing sterile pathways of devices and is used for rinse method testing of medical devices.
Features and Benefits
Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy and reducing the risk of incorrect interpretation and false results.
Analysis Note
The appearance of the medium is clear and colorless to slightly yellowish.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.