Skip to Content
Merck
All Photos(2)

Key Documents

146798

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

Sodium Thiosulfate, Lecithin, Penase, Histidine, Tween®, ICR plus with lockable lid, plate diam. 55 mm, sterile; γ-irradiated, pkg of 10 plates Triple packed, suitable for surface monitoring

Synonym(s):

TSA, Soybean Casein Digest Agar, ICR contact plus plate, Tryptic Soy Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€86.30
200 EA
€859.00

€86.30


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
20 EA
€86.30
200 EA
€859.00

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

€86.30


Estimated to ship on31 March 2025Details


Request more information

material

polystyrene plate

Agency

EP
USP

sterility

sterile; γ-irradiated

form

plate (ready-to-use; safety lock)

feature

ready-to-use

composition

Histidine
Lecithin
Penase
Sodium Thiosulfate
Tween®

packaging

pkg of 10 plates Triple packed
box of 20 ea
box of 200 ea

plate diam.

55 mm

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

shipped in

ambient

storage temp.

2-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar with Penase (Penicillinase) LTHThio in ICR contact plus plate is designed for the determination of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in isolators and clean room. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium and the medium is supplemented with pyruvate to provide an efficient neutralization of hydrogen peroxide residues. Lecithin, polysorbate 80 (Tween®), histidine and sodium thiosulfate are neutralizers to neutralize the disinfectants containing active agents such as Alcohol (70 % ethanol or isopropyl alcohol), Aldehyde, Chlorites, Dichloroisocyanurate, Glucoprotamine, Peracetic acid, Phenols, quaternary ammonium compounds and Biguanides. The medium is also supplemented with penicillinase (≥ 1000 IU/liter) to neutralize residuals of penicillins, such as benzylpenicillin, ampicillin, amoxycillin, carbenicillin and methicillin. In addition, the plate model (plus or “+”) is supplied with a lockable lid for safe transport after sampling.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service