Skip to Content
Merck
All Photos(1)

Key Documents

1.46766

Millipore

ReadyPlate 55 KIT Slanetz&Bartley ISO 7899

for Enterococcus spp., suitable for filtration, box of 150 sets (55 mm plate and EZ-PAK membrane filter)

Sign Into View Organizational & Contract Pricing

Select a Size

150 SETS
€1,240.00

€1,240.00


In StockDetails


Request more information

Select a Size

Change View
150 SETS
€1,240.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€1,240.00


In StockDetails


Request more information

material

gridded white mixed cellulose esters (MCE) membrane

feature

ready-to-use

packaging

box of 150 sets (55 mm plate and EZ-PAK membrane filter)

manufacturer/tradename

ReadyPlate

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

Enterococcus spp.

General description

Application: Detection and enumeration of intestinal enterococci in water using membrane filtration method acc. to ISO

Packaging: 55 mm plate and EZ PAK membrane filter (150 sets per box)

Synonym: m-Enterococcus Agar acc. Slanetz and Bartley, Enterococcus Selective Agar

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service