1.46682
Buffered NaCl Peptone Solution + 5% Tween® 80
bottle capacity 125 mL, bottle filling volume 20 mL, closure type, Blue screw cap with septum and protector, pack of 10 bottles
Synonym(s):
NaCl BPW
About This Item
Recommended Products
Agency
EP 2.6.1
JP 4.06
USP 71
71
Quality Level
sterility
sterile; autoclaved
form
liquid
feature
closure type Blue screw cap with septum and protector
ready-to-use
packaging
pack of 10 bottles
technique(s)
direct inoculation: suitable
microbiological culture: suitable
sterile filtration: suitable
bottle capacity
125 mL
bottle filling volume
20 mL
pH
7.0±0.2
application(s)
cosmetics
food and beverages
pharmaceutical
components
(Tween® 20, Lecithin)
tween: 5%
storage temp.
2-25°C
suitability
nonselective for
General description
Polysorbate 80 or Tween® 80, is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters.
Application
Features and Benefits
- Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
- Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
- Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service