Skip to Content
Merck
All Photos(1)

Key Documents

1.46622

Millipore

ReadyTube® Modified Semi-Solid RAPPAPORT-VASSILIADIS Medium

For the selective enrichment of samples for Salmonella sp. in accordance to ISO 6579, pkg of (12 mL in 17 mL tube with red cap (100 tubes per box))

Synonym(s):

MSRV Semi-solid Medium; Salmonella Enrichment Medium acc. to Rappaport and Vassiliadis

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€200.00

€200.00


Estimated to ship on31 March 2025Details


Request more information

Select a Size

Change View
100 EA
€200.00

About This Item

UNSPSC Code:
41106225
NACRES:
NA.74

€200.00


Estimated to ship on31 March 2025Details


Request more information

Agency

ISO 6579

Quality Level

product line

ReadyTube®

form

semisolid

feature

ready-to-use

packaging

pkg of (12 mL in 17 mL tube with red cap (100 tubes per box))

pH

5.2 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

storage temp.

2-8°C

suitability

selective for Salmonella spp.

General description

ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® modified semi-Solid RAPPAPORT-VASSILIADIS medium is used for the selective enrichment and presumptive detection of Salmonella sp. This medium is a semi-solid modification of Rappaport-Vassiliadis enrichment broth and is used for motility enrichment in Petri dishes. It is a rapid and sensitive medium for the isolation of Salmonella sp. and can be seeded directly after pre-enrichment.

Application

ReadyTube® modified semi-Solid RAPPAPORT-VASSILIADIS medium is used for the selective enrichment and presumptive detection of Salmonella from food, animal feed, and environmental samples from primary production stage

Analysis Note

Peptone and casein hydrolysate provide nitrogenous, carbonaceous, B complex vitamins and other essential growth nutrients. Phosphate provides buffering capacity to the medium. Malachite green serves as a selective agent. The addition of novobiocin as a supplement in the medium selectively inhibits most Gram-positive organisms. Sodium chloride maintains the osmotic equilibrium of the media. Salmonella generally survives a slightly high osmotic pressure (due to MgCl2 in the medium), grows at a slightly low pH, and is resistant to malachite green compared to other bacteria. Its efficiency is due to the ability of Salmonellae to migrate into the semi-solid medium, thus forming opaque halos of growth. All these factors make the medium selective for the isolation of Salmonella. Also, the semi-solid nature of the medium helps to differentiate the motile Salmonella from non-motile ones.

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service