Skip to Content
Merck
All Photos(1)

Key Documents

1.46555

Millipore

Chocolate Agar contact ICR plus

Chocolate agar with Lecithin, Tween® and Histidine- ICR plus Contact plate, irradiated, triple bagged for environmental monitoring (Isolator and Clean room)

Synonym(s):

Chocolate Contact Agar, ICR plus contact plate

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
€96.30

€96.30


Estimated to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
20 PKG
€96.30

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

€96.30


Estimated to ship on31 March 2025Details


Request a Bulk OrderRequest more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate (ready-to-use)

feature

ready-to-use

D

55 mm

pH

7.3 ( in H2O)

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

storage temp.

15-25°C

suitability

bacteria
fungi

General description

Chocolate Contact Agar + LTH – ICR+ is designed for the determination of the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is a universal blood containing culture medium for enrichment and isolation of particularly fastidious bacteria. This medium is characterized by a rich nutritive basis. Dextrose is the source of carbon and energy. In addition, haemin and NAD are released from the sheep blood erythrocytes during heating. Lecithin, Histidine and polysorbate 80 (Tween®) are neutralizers to inactivate potential residues of disinfectants. In addition, ICRplus plates (plus or “+”) is supplied with a lockable lid for safe transport after sampling without the risk of losing the lid as well as for aerobic (closed position) and anaerobic or microaerophilic incubation in the “VENT” position.

Application

Chocolate Contact Agar is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial monitoring; environmental monitoring; interim storage; ICR contact plates; contact plates; TSA w. LTHThio Contact ICR+

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service