Skip to Content
Merck
All Photos(1)

Key Documents

14-654-M

Sigma-Aldrich

eEF-2K Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€3,340.00

€3,340.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 μG
€3,340.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€3,340.00


Estimated to ship on03 March 2025Details


Request a Bulk Order

Quality Level

description

protein target eEF-2K

purified by

(Glutathione-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... EEF2K(29904)

General description

FUNCTION: SwissProt: O00418 # Phosphorylates eukaryotic elongation factor-2. Binds calmodulin (By similarity).
SIZE: 725 amino acids; 82172 Da
SUBUNIT: Monomer or homodimer (Potential).
SIMILARITY: SwissProt: O00418 ## Contains 1 alpha-type protein kinase domain.

Packaging

Also available in 10 μg size (cat#14-654) and in bulk (cat#14-654-K).

Quality

Routinely evaluated by phosphorylation of EF2tide (Catalog# 12-560)
routinely evaluated by phosphorylation of EF2tide (Catalog# 12-560)

Physical properties

108.5kDa

Storage and Stability

stable 6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service