Skip to Content
Merck
All Photos(1)

Key Documents

14-644-M

Sigma-Aldrich

EphA3 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€419.00
250 μG
€3,340.00

€419.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
€419.00
250 μG
€3,340.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.28

€419.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Quality Level

100
300

description

protein target EphA3

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... EPHA3(2042)

General description

N-terminal 6His-tagged, recombinant, human EphA3 amino acids 578-end.

Packaging

Also available in 10 μg size (cat#14-644) and in bulk (cat#14-644-K).

Quality

Routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1)
routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1)

Physical properties

49kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service