Skip to Content
Merck
All Photos(1)

Key Documents

1.46373

Millipore

Mossel Broth (EE Broth)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212

form

liquid

feature

ready-to-use

pH

7.2 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

suitability

enterobacteriaceae

General description

Application: Enrichment of Enterobacteriaceae (Enterobacter)

Packaging: 100 ml in 125 ml bottle with blue screw cap and 3 loci (10 bottles per box)

Synonym: Buffered Glucose-Brilliant Green Bile Broth, EE Broth, Enterobacteriaceae Enrichment Broth acc. to Mossel

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service