Skip to Content
Merck
All Photos(2)

Key Documents

1.46283

Millipore

Tryptic Soy Broth- Ready-to-use Bags, Tubes & Ampoules

Histidine, Lecithin, Tween®, Thiosulfate, tube capacity × filling volume 17 mL × 10 mL, closure type, blue screw cap with 2 loci, box of 20 or 100 tubes

Synonym(s):

CASO, Casein Soybean Digest Broth, Casein Soybean Digest Broth with Lecithin, Tween, Histidine, and Thiosulfate, SCDB, TSB with neutralizer, Tryptic Soy Broth, Tryptic Soy Broth with TLHTh

Sign Into View Organizational & Contract Pricing

Select a Size

20 TUBES
€40.70
100 TUBES
€269.00

€40.70


Estimated to ship on24 March 2025Details


Request more information

Select a Size

Change View
20 TUBES
€40.70
100 TUBES
€269.00

About This Item

UNSPSC Code:
41106226
NACRES:
NA.74

€40.70


Estimated to ship on24 March 2025Details


Request more information

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

description

with Lecithin, Tween®, Histidin and Thiosulfate, for the isolation and cultivation of fastidious aerobic bacteria, yeasts, & molds; Suitable for sterility testing

sterility

sterile; autoclaved

form

liquid (ready-to-use)

shelf life

12 mo.

feature

closure type blue screw cap with 2 loci
ready-to-use

composition

Histidine
Lecithin
Thiosulfate
Tween®

packaging

box of 20 or 100 tubes
tube of

capacity

17 mL

tube capacity × filling volume

17 mL × 10 mL

color

clear yellowish to yellow

pH

7.3

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Tryptic Soy Broth with neutralizers is a universal complex and highly nutritious medium and supports the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi. The neutralizing additives Polysorbate (Tween®) 80, sodium thiosulfate, cysteine, histidine, and lecithin reverse the growth inhibitory effect of most disinfectants and antiseptics. Histidine neutralizes formaldehyde and formaldehyde releasing agents. Sodium thiosulfate inactivates sodium hypochlorite and acidified sodium chlorite. Cysteine neutralizes the oxidizing agents, such as hydrogen peroxide and iodine. Lecithin along with Polysorbate 80 is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrols (PHB esters).

Application

Tryptic Soy Broth with LTHTh is used for the isolation and cultivation of fastidious aerobic bacteria, yeasts and molds in the presence of disinfectant residues. The medium designed for the testing of sterile and non-sterile substances, preparations and products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service